2-Acetyl-4-chlorothiophene ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 1-(4-Chlorothiophen-2-yl)ethanone, 1-(4-Chloro-2-thienyl)-1-ethanone

H30384.06 H30384.14
H30384.06EA 114 EUR
H30384.06 H30384.14
2-Acetyl-4-chlorothiophene ≥98%
2-Acetyl-4-chlorothiophene

Purity: 98+% CAS No.: 34730-20-6 Molecular Formula: C6H5ClOS Formula Weight: 160.62 UN No.: 2810 MDL No.: MFCD00082791

Formula: C₆H₅ClOS
Density: 1,334
Storage Temperature: Ambient
MDL Number: MFCD00082791
CAS Number: 34730-20-6
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR