5-Amino-2-fluoropyridine ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Fluoro-5-pyridylamine, 2-Fluoropyridin-5-amine

H30872.03
H30872.03EA 215 EUR
H30872.03
5-Amino-2-fluoropyridine ≥97%
5-Amino-2-fluoropyridine

Purity: 97+% CAS No.: 1827-27-6 Molecular Formula: C5H5FN2 Formula Weight: 112.11 Odour: Characteristic Melting Point: 88-91° UN No.: 2811 MDL No.: MFCD01632180

Formula: C₅H₅FN₂
MW: 112,11 g/mol
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD01632180
CAS Number: 1827-27-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR