2-Phenylquinoline ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
H31920.03
H31920.06
H31920.03EA
86.8
EUR
H31920.03
H31920.06
2-Phenylquinoline ≥99%
2-Phenylquinoline
Purity: 99+%
CAS No.: 612-96-4
Molecular Formula: C15H11N
Formula Weight: 205.26
Melting Point: 82-84°
Boiling Point: 363°
MDL No.: MFCD00011568
Formula:
C₁₅H₁₁N MW: 205.26 g/mol Boiling Pt: 363 °C Melting Pt: 82…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011568 CAS Number: 612-96-4 EINECS: 210-326-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...