3-Fluoro-5-hydroxypyridine ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Fluoro-5-pyridinol, 5-Fluoropyridin-3-ol
H32683.MD
H32683.03
H32683.MDEA
98.7
EUR
H32683.MD
H32683.03
3-Fluoro-5-hydroxypyridine ≥97%
3-Fluoro-5-hydroxypyridine
Purity: 97%
CAS No.: 209328-55-2
Molecular Formula: C5H4FNO
Formula Weight: 113.09
Melting Point: 161-164°
MDL No.: MFCD03095199
Formula:
C₅H₄FNO MW: 113,09 g/mol Melting Pt: 161…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095199 CAS Number: 209328-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...