4-(1-Aminoethyl)pyridine 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
1-Pyridin-4-ylethylamine, 1-(4-Pyridyl)ethylamine
H50303.03
H50303.MD
H50303.03EA
114
EUR
H50303.03
H50303.MD
4-(1-Aminoethyl)pyridine 97%
4-(1-Aminoethyl)pyridine
Purity: 97%
CAS No.: 50392-78-4
Molecular Formula: C7H10N2
Formula Weight: 122.17
UN No.: 2735
MDL No.: MFCD02245004
Formula:
C₇H₁₀N₂ MW: 122,17 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD02245004 CAS Number: 50392-78-4 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...