N-(3-Bromophenyl)maleimide 96%

Supplier: Thermo Scientific
Warning

H50586.03 H50586.MD
H50586.03EA 105 EUR
H50586.03 H50586.MD
N-(3-Bromophenyl)maleimide 96%
N-(3-Bromophenyl)maleimide

Purity: 96% CAS No.: 53534-14-8 Molecular Formula: C10H6BrNO2 Formula Weight: 252.07 MDL No.: MFCD01026470

Formula: C₁₀H₆BrNO₂
MW: 252,07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01026470
CAS Number: 53534-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR