Allyltrichlorosilane 95%

Supplier: Thermo Fisher Scientific
Danger

L03703.06 L03703.14
L03703.06EA 65.4 EUR
L03703.06 L03703.14
Allyltrichlorosilane 95%
Allyltrichlorosilane

Purity: 95% CAS No.: 107-37-9 Molecular Formula: C3H5Cl3Si Formula Weight: 175.52 Flash Point: 18°(64°F) Refractive Index: 1.4490 Boiling Point: 117-118° Beilstein Reference: 1743449 UN No.: 1724 MDL No.: MFCD00000486

Formula: H₂CCHCH₂SiCl₃
Boiling Pt: 117…118 °C
Melting Pt: 35 °C
Density: 1,211
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000486
CAS Number: 107-37-9
EINECS: 203-485-9
UN: 1724
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR