Ethyl chloroformate ≥97%

Supplier: Thermo Scientific
Danger

Synonyms: Chloroformic acid ethyl ester

L06311.22 L06311.36 L06311.06 L06311.C4 L06311.PG
L06311.22EA 36.3 EUR
L06311.22 L06311.36 L06311.06 L06311.C4 L06311.PG
Ethyl chloroformate ≥97%
Ethyl chloroformate
Formula: C₃H₅ClO₂
MW: 108,52 g/mol
Boiling Pt: 92…93 °C
Melting Pt: –81 °C
Density: 1,136
Flash Pt: 10 °C (50 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000644
CAS Number: 541-41-3
EINECS: 208-778-5
UN: 1182
ADR: 6.1,I
Merck Index: 14,3784

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR