3-Cyanophenyl isocyanate ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Cyanophenylisocyanate, 3-Isocyanatobenzonitrile

L09432.03 L09432.06
L09432.03EA 36.7 EUR
L09432.03 L09432.06
3-Cyanophenyl isocyanate ≥97%
3-Cyanophenyl isocyanate

Purity: 97% CAS No.: 16413-26-6 Molecular Formula: C8H4N2O Formula Weight: 144.13 Storage Temperature: 2°C to 8°C Odour: Irritating Beilstein Reference: 3089117 UN No.: 2811 MDL No.: MFCD00013856

Formula: C₈H₄N₂O
MW: 144,13 g/mol
Boiling Pt: 123 °C (12 mmHg)
Melting Pt: 51…54 °C
MDL Number: MFCD00013856
CAS Number: 16413-26-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR