4-Bromo-2-methylphenyl isocyanate ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Bromo-2-methylphenylisocyanate
L11095.03
L11095.06
L11095.03EA
49.8
EUR
L11095.03
L11095.06
4-Bromo-2-methylphenyl isocyanate ≥98%
4-Bromo-2-methylphenyl isocyanate
Purity: 98%
CAS No.: 1591-98-6
Molecular Formula: C8H6BrNO
Formula Weight: 212.05
Storage Temperature: 2°C to 8°C
Flash Point:
Formula:
C₈H₆BrNO MW: 212,05 g/mol Boiling Pt: 265 °C (1013 hPa) Melting Pt: 47…51 °C Flash Pt: >110 °C (230 °F) |
MDL Number:
MFCD00037039 CAS Number: 1591-98-6 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...