(3,5-Dimethoxyphenyl)acetonitrile ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 3,5-Dimethoxyphenylacetonitrile, 3,5-Dimethoxybenzyl cyanide, 2-(3,5-Dimethoxyphenyl)acetonitrile

L11443.03 L11443.06
L11443.03EA 66.8 EUR
L11443.03 L11443.06
(3,5-Dimethoxyphenyl)acetonitrile ≥98%
(3,5-Dimethoxyphenyl)acetonitrile

Purity: 98+% CAS No.: 13388-75-5 Molecular Formula: C10H11NO2 Formula Weight: 177.20 Odour: Characteristic Melting Point: 53-56° Beilstein Reference: 2096481 MDL No.: MFCD00016395

Formula: C₁₀H₁₁NO₂
MW: 177,2 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 53…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00016395
CAS Number: 13388-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR