3-Methylbenzyl mercaptan ≥95%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
m-Xylene-α-thiol, 3-Methylbenzylmercaptan
L12043.06
L12043.03
L12043.06EA
0
EUR
L12043.06
L12043.03
3-Methylbenzyl mercaptan ≥95%
3-Methylbenzyl mercaptan
Purity: 95%
CAS No.: 25697-56-7
Molecular Formula: C8H10S
Formula Weight: 138.23
Refractive Index: 1.5630
Boiling Point: 102-104°/15mm
UN No.: 2810
MDL No.: MFCD00022075
Formula:
C₈H₁₀S MW: 138,23 g/mol Boiling Pt: 102…104 °C (15 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00022075 CAS Number: 25697-56-7 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...