3,5-Di-tert-butylbenzoic acid ≥99%
Supplier: Thermo Fisher Scientific
|
Warning
|
L12386.03
L12386.06
L12386.03EA
23
EUR
L12386.03
L12386.06
3,5-Di-tert-butylbenzoic acid ≥99%
3,5-Di-tert-butylbenzoic acid
Purity: 99%
CAS No.: 16225-26-6
Molecular Formula: C15H22O2
Formula Weight: 234.34
Melting Point: 160-162°
Beilstein Reference: 981248
Formula:
C₁₅H₂₂O₂ MW: 234.34 g/mol Melting Pt: 160…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082727 CAS Number: 16225-26-6 EINECS: 240-350-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...