Trifluoroacetamidine tech. 85%, Technical Grade
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2,2,2-Trifluoroacetimidamide, 2,2,2-Trifluoroacetamidine
L12422.06
L12422.14
L12422.22
L12422.06EA
108
EUR
L12422.06
L12422.14
L12422.22
Trifluoroacetamidine tech. 85%, Technical Grade
Trifluoroacetamidine
Purity: 85%
CAS No.: 354-37-0
Molecular Formula: C2H3F3N2
Formula Weight: 112.05
Storage Temperature: -30°C to -10°C
Refractive Index: 1.3820
Boiling Point: 35-37°/11mm
Beilstein Reference: 605978
UN No.: 3267
MDL No.: MFCD00041545
Formula:
C₂H₃F₃N₂ MW: 112,05 g/mol Boiling Pt: 35…37 °C (11 mmHg) Density: 1,45 |
MDL Number:
MFCD00041545 CAS Number: 354-37-0 UN: 3267 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...