Hept-6-enoic acid 96%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98%
CAS No.: 1119-60-4
Molecular Formula: CH2=CH(CH2)4CO2H
Formula Weight: 128.17
Flash Point:
Formula:
C₇H₁₂O₂ Boiling Pt: 222…224 °C Melting Pt: ∼–6 °C Density: 0,946 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014382 CAS Number: 1119-60-4 EINECS: 214-283-5 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...