n-Butyllithium 1.6 mol/l in hexane
Supplier: Thermo Scientific
L13559.AE
L13559.AP
L13559.AEEA
73.5
EUR
L13559.AE
L13559.AP
n-Butyllithium 1.6 mol/l in hexane
n-Butyllithium
Formula:
C₄H₉Li MW: 64,06 g/mol Density: 0,69 g/cm³ (20 °C) Flash Pt: –22 °C |
MDL Number:
MFCD00009414 CAS Number: 109-72-8 EINECS: 203-698-7 UN: 3394 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...