3-Chloro-2-thenoic acid ≥99%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
3-Chloro-2-thiophenecarboxylic acid, 3-Chlorothiophene-2-carboxylic acid
L13560.03
L13560.06
L13560.03EA
49.7
EUR
L13560.03
L13560.06
3-Chloro-2-thenoic acid ≥99%
3-Chloro-2-thenoic acid
Purity: 99%
CAS No.: 59337-89-2
Molecular Formula: C5H3ClO2S
Formula Weight: 162.60
Melting Point: 185-187°
Beilstein Reference: 121052
Formula:
C₅H₃ClO₂S MW: 162,6 g/mol Melting Pt: 185…187 °C Storage Temperature: Ambient |
MDL Number:
MFCD00043888 CAS Number: 59337-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...