Isoprene ≥99% stabilised
Supplier: Thermo Fisher Scientific
|
Danger
|
Stabilisation:
Stabilised with 4-tert-butylpyrocatechol (TBC) 200 ppm
L14619.AE
L14619.AP
L14619.AEEA
28.4
EUR
L14619.AE
L14619.AP
Isoprene ≥99% stabilised
Isoprene
Stabilized with ca 0.02% 4-tert-butylcatechol
Purity: 99%
CAS No.: 78-79-5
Molecular Formula: C5H8
Formula Weight: 68.12
Storage Temperature: 2°C to 8°C
Flash Point: -54°(-65°F)
Refractive Index: 1.4220
Melting Point: -146°
Boiling Point: 33-36°
Beilstein Reference: 969158
Merck Reference: 14,5201
UN No.: 1218
MDL No.: MFCD00008600
Formula:
C₅H₈ Boiling Pt: 33…36 °C Melting Pt: –146 °C Density: 0,681 Flash Pt: –54 °C (–65 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008600 CAS Number: 78-79-5 EINECS: 201-143-3 UN: 1218 ADR: 3,I Merck Index: 14,5201 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...