5-(2,4-Dichlorophenyl)-1H-tetrazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
L15642.03
L15642.06
L15642.MD
L15642.03EA
84.4
EUR
L15642.03
L15642.06
L15642.MD
5-(2,4-Dichlorophenyl)-1H-tetrazole ≥97%
5-(2,4-Dichlorophenyl)-1H-tetrazole
Purity: 97%
CAS No.: 50907-22-7
Molecular Formula: C7H4Cl2N4
Formula Weight: 215.04
Melting Point: 162-164°
MDL No.: MFCD01075709
Formula:
C₇H₄Cl₂N₄ MW: 215,04 g/mol Melting Pt: 162…164 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075709 CAS Number: 50907-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...