5-(3,4-Dichlorophenyl)-1H-tetrazole ≥97%

Supplier: Thermo Scientific
Warning

L15643.03 L15643.MD
L15643.03EA 152 EUR
L15643.03 L15643.MD
5-(3,4-Dichlorophenyl)-1H-tetrazole ≥97%
5-(3,4-Dichlorophenyl)-1H-tetrazole

Purity: 97% CAS No.: 41421-27-6 Molecular Formula: C7H4Cl2N4 Formula Weight: 215.04 Melting Point: 158-163° MDL No.: MFCD01075710

Formula: C₇H₄Cl₂N₄
MW: 215,04 g/mol
Melting Pt: 158…163 °C
Storage Temperature: Ambient
MDL Number: MFCD01075710
CAS Number: 41421-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR