3,5-Difluorobenzeneboronic acid neopentyl glycol ester ≥98%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-(3,5-Difluorophenyl)-5,5-dimethyl-1,3,2-dioxaborinane

L17266.03 L17266.06
L17266.03EA 0 EUR
L17266.03 L17266.06
3,5-Difluorobenzeneboronic acid neopentyl glycol ester ≥98%
3,5-Difluorobenzeneboronic acid neopentyl glycol ester
Formula: C₆H₅BF₂O₂
Melting Pt: 43…45 °C
Storage Temperature: Ambient
MDL Number: MFCD01318138
CAS Number: 156545-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR