2-Chloro-6-(trifluoromethyl)pyridine ≥97%

Supplier: Thermo Scientific
Danger

L20034.03 L20034.06
L20034.03EA 0 EUR
L20034.03 L20034.06
2-Chloro-6-(trifluoromethyl)pyridine ≥97%
2-Chloro-6-(trifluoromethyl)pyridine

Purity: 97% CAS No.: 39890-95-4 Molecular Formula: C6H3ClF3N Formula Weight: 181.55 Flash Point: 78°(172°F) Melting Point: 28-29° Boiling Point: 60-62°/8mm Beilstein Reference: 4308910 UN No.: 2811 MDL No.: MFCD00042226

Formula: C₆H₃ClF₃N
MW: 181,54 g/mol
Boiling Pt: 60…62 °C (8 mmHg)
Melting Pt: 28…29 °C
Flash Pt: 78 °C (172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042226
CAS Number: 39890-95-4
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR