Phthalaldehyde, Millipore®
Supplier: Merck
|
Danger
|
Synonyms:
1,2-Phthalic dicarboxaldehyde, Phthaldialdehyde, o-Phthalaldehyde, OPA, Phthaldicarboxaldehyde, Benzene-1,2-dicarboxaldehyde
M-Clarity™ quality level = MQ100
1.11452.0005
1.11452.0050
1.11452.0005EA
101
EUR
1.11452.0005
1.11452.0050
Phthalaldehyde, Millipore®
Phthalaldehyde
Formula:
C₈H₆O₂ MW: 134,13 g/mol Boiling Pt: 266 °C (1013 hPa) Melting Pt: 54…56 °C Density: 1,13 g/cm³ (20 °C) Flash Pt: 132 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003335 CAS Number: 643-79-8 EINECS: 211-402-2 UN: 2923 ADR: 8 (6.1), |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...