2-Aminothiazole, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Thiazol-2-ylamine, Aminothiazole, Thiazol-2-amine
M-Clarity™ quality level = MQ200
8.01272.0100
8.01272.0500
8.01272.0100EA
37.5
EUR
8.01272.0100
8.01272.0500
2-Aminothiazole, Sigma-Aldrich®
2-Aminothiazole
Formula:
C₃H₄N₂S MW: 100.14 g/mol Boiling Pt: 216 Melting Pt: 86…89 °C Density: 1.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005325 CAS Number: 96-50-4 EINECS: 202-511-6 Merck Index: 13,00479 |
Specification Test Results
Solubility | 100 g/l (20 °C) |
Melting Point | 86 - 89 °C |
Molar Mass | 100.13 g/mol |
Bulk Density | 600 kg/m3 |
Boiling Point | 117 °C (20 hPa) |
Vapor Pressure |
|
pH | 9.6 (100 g/l, H2O, 20 °C) |
Ignition Point | 600 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...