Trichloromethyl chloroformate, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Chloroformic acid trichloromethyl ester, Diphosgene
M-Clarity™ quality level = MQ200
8.18154.0025
8.18154.0100
8.18154.0025EA
234
EUR
8.18154.0025
8.18154.0100
Trichloromethyl chloroformate, Sigma-Aldrich®
Trichloromethyl chloroformate
Formula:
C₂Cl₄O₂ MW: 197,83 g/mol Boiling Pt: 128 °C (1013 hPa) Melting Pt: –57 °C Density: 1,63 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00015553 CAS Number: 503-38-8 EINECS: 207-965-9 UN: 3389 ADR: 6.1 (8),I Merck Index: 13,03368 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...