Isoprene stabilised, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.21258.0100 8.21258.1000
8.21258.0100EA 34.3 EUR
8.21258.0100 8.21258.1000
Isoprene stabilised, Sigma-Aldrich®
Isoprene
Formula: C₅H₈
Boiling Pt: 34 °C (1013 hPa)
Melting Pt: –146 °C
Density: 0,681 g/cm³ (20 °C)
Flash Pt: –48 °C
MDL Number: MFCD00008600
CAS Number: 78-79-5
EINECS: 201-143-3
UN: 1218
ADR: 3,I
Merck Index: 13,05220

Order Now

Specification Test Results

Solubility 0.7 g/l (20 °C)
Melting Point -146 °C
Molar Mass 68.12 g/mol
Boiling Point 34 °C (1013 hPa)
Vapor Pressure 605 hPa (20 °C)
Flash Point -54 °C
Refractive Index 1.4217 (20 °C, 589 nm)
Explosion Limit 1 - 9.7 %(V)
Density 0.68 g/cm3 (20 °C)
Ignition Point 220 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR