(Chloromethyl)trichlorosilane, Sigma-Aldrich®

Supplier: Merck
Danger


M-Clarity™ quality level = MQ200

8.42025.0005 8.42025.0025
8.42025.0005EA 63.7 EUR
8.42025.0005 8.42025.0025
(Chloromethyl)trichlorosilane, Sigma-Aldrich®
(Chloromethyl)trichlorosilane
Formula: ClH₂CSiCl₃
Boiling Pt: 117 °C (1013 hPa)
Density: 1,442 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000873
CAS Number: 1558-25-4
EINECS: 216-316-9
UN: 3389
ADR: 6.1 (8),I

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR