Periodic acid
Supplier: Roth Carl
|
Danger
|
Synonyms:
ortho-Periodic acid, Orthoperiodic acid
3257.1
3257.2
ROTH3257.1EA
0
EUR
ROTH3257.1
ROTH3257.2
Periodic acid
Periodic acid
Formula:
H₅IO₆ MW: 227.94 g/mol Melting Pt: 122 Density: 1.3875 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011344 CAS Number: 10450-60-9 EINECS: 233-937-0 UN: 3085 ADR: 5.1 I Merck Index: 13,07248 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...