Levulinic acid
Supplier: Roth Carl
Synonyms:
4-Oxovaleric acid, 4-Ketovaleric acid, 4-Oxopentanoic acid
3976.1
3976.2
ROTH3976.1EA
22.2
EUR
ROTH3976.1
ROTH3976.2
Levulinic acid
Levulinic acid
Formula:
C₅H₈O₃ Boiling Pt: 245 °C (1013 hPa) Melting Pt: 26…29 °C Density: 1,14 g/cm³ (20 °C) Flash Pt: 138 °C |
MDL Number:
MFCD00002796 CAS Number: 123-76-2 EINECS: 204-649-2 UN: 3261 ADR: 8,III Merck Index: 13,05492 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...