N-Phenylethylenediamine 99%
Supplier: Thermo Fisher Scientific
|
Danger
|
130610100
130610500
ACRO130610100EA
69
EUR
ACRO130610100
ACRO130610500
N-Phenylethylenediamine 99%
N-Phenylethylenediamine
Formula:
C₆H₅NHCH₂CH₂NH₂ MW: 136.2 g/mol Boiling Pt: 262…264 °C (760 mmHg) Density: 1.04 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008162 CAS Number: 1664-40-0 EINECS: 216-773-4 UN: 2735 ADR: 8 III |
Specification Test Results
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5855 to 1.5875 (20°C, 589 nm) |
Titration with HClO4 | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...