(4-Chlorophenyl)acetyl chloride 96%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-(4-Chlorophenyl)acetyl chloride, 4-Chlorobenzeneacetyl chloride
123450050
123450250
123451000
ACRO123450050EA
35.3
EUR
ACRO123450050
ACRO123450250
ACRO123451000
(4-Chlorophenyl)acetyl chloride 96%
(4-Chlorophenyl)acetyl chloride
Formula:
(C₆H₄Cl)CH₂COCl MW: 189,04 g/mol Boiling Pt: 85 °C (1 mmHg) Density: 1,292 g/cm³ Flash Pt: >110 °C |
MDL Number:
MFCD00037111 CAS Number: 25026-34-0 EINECS: 246-571-1 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear yellow to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5490 to 1.5570 (20°C, 589 nm) |
GC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...