4-Iodo-3,5-xylenol ≥98%
Supplier: Thermo Scientific
|
Warning
|
A17292.06
A17292.03
A17292.06EA
0
EUR
A17292.06
A17292.03
4-Iodo-3,5-xylenol ≥98%
4-Iodo-3,5-xylenol
Purity: 98%
CAS No.: 80826-86-4
Molecular Formula: C8H9IO
Formula Weight: 248.06
Melting Point: 129-133°
MDL No.: MFCD00174284
Formula:
C₈H₉IO Melting Pt: 129…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD00174284 CAS Number: 80826-86-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...