4-Chloro-2-methoxybenzyl alcohol ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: (4-Chloro-2-methoxyphenyl)methanol

H27562.14
H27562.14EA 362 EUR
H27562.14
4-Chloro-2-methoxybenzyl alcohol ≥97%
4-Chloro-2-methoxybenzyl alcohol

Purity: 97% CAS No.: 55685-75-1 Molecular Formula: C8H9ClO2 Formula Weight: 172.61 Flash Point: 110°(230°F) Melting Point: 46-50° MDL No.: MFCD02683547

Formula: C₈H₉ClO₂
MW: 172,61 g/mol
Melting Pt: 46…50 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD02683547
CAS Number: 55685-75-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR