2-Acetylphenylboronic acid 96%

Supplier: Thermo Scientific
Warning

Synonyms: (2-Acetylphenyl)metaboric acid, 2-Acetylbenzeneboronic acid, (2-Acetylphenyl)boronic acid, Dihydroxy(2-acetylphenyl)borane

358820010 358820050 358820250
ACRO358820010EA 64.7 EUR
ACRO358820010 ACRO358820050 ACRO358820250
2-Acetylphenylboronic acid 96%
2-Acetylphenylboronic acid
Formula: C₈H₉BO₃
MW: 163,97 g/mol
Melting Pt: 170 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD01321263
CAS Number: 308103-40-4

Order Now

Specification Test Results

Appearance White to yellow Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR