(±)-Citronellal 93% pract.

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (+/-)-Citronellal, (±)-3,7-Dimethyloct-6-enal, (+/-)-3,7-Dimethyl-6-octenal

405291000 405295000
ACRO405291000EA 40.3 EUR
ACRO405291000 ACRO405295000
(±)-Citronellal 93% pract.
(±)-Citronellal
Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 207
Melting Pt: –15 °C
Density: 0.86 g/cm³
Flash Pt: 78 °C
MDL Number: MFCD00038090
CAS Number: 106-23-0
EINECS: 203-376-6
Merck Index: 13,02353

Order Now

Specification Test Results

Appearance to light green-yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4500 (20°C, 589 nm)
GC ≥91 %
Specific gravity 0.85 to 0.87 g/ml (20°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR