4-Chloro-2-fluorophenylboronic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Chloro-2-fluorobenzeneboronic acid, (4-Chloro-2-fluorophenyl)metaboric acid, Dihydroxy(4-chloro-2-fluorophenyl)borane

H28872.09
H28872.09EA 222 EUR
H28872.09
4-Chloro-2-fluorophenylboronic acid ≥97%
4-Chloro-2-fluorophenylboronic acid

Purity: 97% CAS No.: 160591-91-3 Molecular Formula: C6H5BClFO2 Formula Weight: 174.37 Melting Point: 246-250° MDL No.: MFCD02684293

Formula: C₆H₅BClFO₂
MW: 174,37 g/mol
Melting Pt: 246…250 °C
Storage Temperature: Ambient
MDL Number: MFCD02684293
CAS Number: 160591-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR