α-Angelicalactone ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Methylfuran-2(3H)-one

L07602.06 L07602.14 L07602.22
L07602.06EA 30.7 EUR
L07602.06 L07602.14 L07602.22
α-Angelicalactone ≥98%
α-Angelicalactone

Purity: 98% CAS No.: 591-12-8 Molecular Formula: C5H6O2 Formula Weight: 98.10 Flash Point: 68°(154°F) Refractive Index: 1.4480 Melting Point: 17-18° Boiling Point: 53-54°/10mm Beilstein Reference: 108394 Merck Reference: 14,647 MDL No.: MFCD00005375

Formula: C₅H₆O₂
MW: 98,1 g/mol
Boiling Pt: 53…54 °C (10 mmHg)
Melting Pt: 17…18 °C
Density: 1,093
Flash Pt: 68 °C (154 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005375
CAS Number: 591-12-8
EINECS: 209-701-8
Merck Index: 14,647

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR