2,5-Diaminopyridine dihydrochloride 97%
Supplier: Thermo Scientific
|
Warning
|
L05599.06
L05599.14
L05599.06EA
55.4
EUR
L05599.06
L05599.14
2,5-Diaminopyridine dihydrochloride 97%
2,5-Diaminopyridine dihydrochloride
Purity: 97%
CAS No.: 26878-35-3
Molecular Formula: C5H7N3.2HCl
Formula Weight: 182.05
Melting Point: ca 264° dec.
Beilstein Reference: 3912365
Formula:
C₅H₉Cl₂N₃ MW: 182,05 g/mol Melting Pt: ∼ 264 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00040818 CAS Number: 26878-35-3 EINECS: 248-083-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...