2-Chloro-3-pyridineboronic acid 96%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
2-Chloro-3-pyridinylboronic acid, (2-Chloropyridin-3-yl)boronic acid, 2-Chloropyridine-3-boronic acid
L20303.03
L20303.06
L20303.03EA
42.1
EUR
L20303.03
L20303.06
2-Chloro-3-pyridineboronic acid 96%
2-Chloro-3-pyridineboronic acid
Purity: 96%
CAS No.: 381248-04-0
Molecular Formula: C5H5BClNO2
Formula Weight: 157.37
Melting Point: 160°
Beilstein Reference: 9119913
Formula:
C₅H₅BClNO₂ Melting Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094997 CAS Number: 381248-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...