(±)-3-Hydroxybutyric acid, tech.

Supplier: Thermo Scientific
Warning

Synonyms: 3-Hydroxybutyric acid, DL-3-Hydroxybutyric Acid, (±)-3-Hydroxybutanoic acid

43354.14 43354.06
43354.14EA 384 EUR
43354.14 43354.06
(±)-3-Hydroxybutyric acid, tech.
(±)-3-Hydroxybutyric acid

CAS No.: 300-85-6 Molecular Formula: C4H8O3 Formula Weight: 104.10 Flash Point: 112°(234°F) Physical Form: Liquid Density: 1.13 Refractive Index: 1.4445 Sensitivity: Hygroscopic Appearance: Colourless Boiling Point: 130°/12mm Solubility: Soluble in water, alcohol, ether Merck Reference: 14,4816 MDL No.: MFCD00004546

Formula: C₄H₈O₃
MW: 104,11 g/mol
Boiling Pt: 130 °C (12 mmHg)
Melting Pt: 33…34 °C
Density: 1,13
Flash Pt: >112 °C (234 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004546
CAS Number: 300-85-6
EINECS: 206-099-9
Merck Index: 14,4816

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR