Anti-Mouse IgG Goat Polyclonal Antibody (CF®583R)

Supplier: Biotium
20914-50UL 20914-250UL
20914-50ULEA 103 EUR
20914-50UL 20914-250UL
Anti-Mouse IgG Goat Polyclonal Antibody (CF®583R)
Antibodies
This is a goat anti-mouse IgG (H+L), F(ab’)2 fragment secondary antibody labeled with our superior CF® Dyes and other labels.

  • Available in 14 bright and photostable CF® Dyes, and HRP
  • Biotin, APC, and R-PE conjugates also available
  • Suitable for western, immunofluorescence, and immunohistology in FFPE tissues

Conjugates of blue fluorescent dyes like CF®350 are not recommended for detecting low abundance targets, because blue dyes have lower fluorescence and can give higher non-specific background than other dye colours.

Type: Secondary
Antigen: Mouse IgG
Clonality: Polyclonal
Clone:
Conjugation: CF®583R
Epitope:
Host: Goat
Isotype: IgG (H+L)
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR