3-Bromostyrene ≥97% stabilised
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1-Bromo-3-vinylbenzene
Stabilisation:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0,10 %
B24497.03
B24497.06
B24497.14
B24497.03EA
41.5
EUR
B24497.03
B24497.06
B24497.14
3-Bromostyrene ≥97% stabilised
3-Bromostyrene
Stabilized with 0.1% 4-tert-butylcatechol
Purity: 97%
CAS No.: 2039-86-3
Molecular Formula: C8H7Br
Formula Weight: 183.05
Storage Temperature: 2°C to 8°C
Flash Point: 67°(152°F)
Refractive Index: 1.5910
Boiling Point: 73-74°/3mm
Beilstein Reference: 2038490
MDL No.: MFCD00000088
Formula:
C₈H₇Br MW: 183,05 g/mol Boiling Pt: 73…74 °C (3 mmHg) Density: 1,404 Flash Pt: 67 °C (152 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000088 CAS Number: 2039-86-3 EINECS: 218-025-2 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...