4-Chlorostyrene ≥99% stabilised

Supplier: Thermo Fisher Scientific
Warning


Stabilisation: Stabilised with 4-tert-butylpyrocatechol (TBC)

B20512.09 B20512.18
B20512.09EA 58.2 EUR
B20512.09 B20512.18
4-Chlorostyrene ≥99% stabilised
4-Chlorostyrene

Stabilized


Purity: 99% CAS No.: 1073-67-2 Molecular Formula: C8H7Cl Formula Weight: 138.60 Storage Temperature: 2°C to 8°C Flash Point: 67°(153°F) Refractive Index: 1.5650 Odour: Characteristic Melting Point: -16° Boiling Point: 191-192° Beilstein Reference: 1098859 MDL No.: MFCD00000632

Formula: C₈H₇Cl
Boiling Pt: 191…192 °C
Melting Pt: –16 °C
Density: 1,089
Flash Pt: 67 °C (153 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000632
CAS Number: 1073-67-2
EINECS: 214-028-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR