2-Methylstyrene ≥98% stabilised
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-Vinyltoluene, 1-Methyl-2-vinylbenzene
Stabilisation:
Stabilised with 4-tert-butylpyrocatechol (TBC) 0,10 %
A12676.06
A12676.14
A12676.06EA
88.4
EUR
A12676.06
A12676.14
2-Methylstyrene ≥98% stabilised
2-Methylstyrene
Stabilized with 0.1% 4-tert-butylcatechol
Purity: 98%
CAS No.: 611-15-4
Molecular Formula: C9H10
Formula Weight: 118.18
Storage Temperature: 2°C to 8°C
Flash Point: 58°(136°F)
Refractive Index: 1.5450
Melting Point: -69°
Boiling Point: 170-172°
Beilstein Reference: 1901764
UN No.: 2618
MDL No.: MFCD00014936
Formula:
C₉H₁₀ MW: 118,18 g/mol Boiling Pt: 170…172 °C Melting Pt: –69 °C Density: 0,916 Flash Pt: 58 °C (136 °F) |
MDL Number:
MFCD00014936 CAS Number: 611-15-4 EINECS: 210-256-7 UN: 2618 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...