4-Fluorostyrene ≥97% stabilised

Supplier: Thermo Fisher Scientific
Warning


Stabilisation: Stabilised with 4-tert-butylpyrocatechol (TBC) 0,10 %

A14775.03 A14775.06 A14775.14 A14775.22
A14775.03EA 39.6 EUR
A14775.03 A14775.06 A14775.14 A14775.22
4-Fluorostyrene ≥97% stabilised
4-Fluorostyrene

Stabilized with 0.1% 4-tert-butylcatechol


Purity: 97+% CAS No.: 405-99-2 Molecular Formula: C8H7F Formula Weight: 122.14 Storage Temperature: 2°C to 8°C Flash Point: 26°(78°F) Refractive Index: 1.5150 Odour: Recognizable Melting Point: -36° Boiling Point: 145-148° Beilstein Reference: 1634203 UN No.: 1993 MDL No.: MFCD00000361

Formula: C₈H₇F
Boiling Pt: 145…148 °C
Melting Pt: –36 °C
Density: 1,024
Flash Pt: 26 °C (78 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000361
CAS Number: 405-99-2
EINECS: 206-975-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR