4-Methoxystyrene 96% Stabilised
Supplier: Thermo Fisher Scientific
Synonyms:
1-Methoxy-4-vinylbenzene, 4-Vinylanisole
161390050
161390250
ACRO161390050EA
91.8
EUR
ACRO161390050
ACRO161390250
4-Methoxystyrene 96% Stabilised
4-Methoxystyrene
Formula:
C₉H₁₀O MW: 134.18 g/mol Boiling Pt: 204…205 °C Melting Pt: 3.3 Density: 1 g/cm³ Flash Pt: 76 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008619 CAS Number: 637-69-4 EINECS: 211-298-9 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5600 to 1.5620 (20°C, 589 nm) |
GC | ≥95 % |
Stabilizer | 0.1 % 4-tert-Butylcatechol |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...