4-Methylstyrene 98% stabilised

Supplier: Thermo Scientific
Danger

Synonyms: p-Vinyltoluene, p-Methylstyrene, 4-Vinyltoluene

127700100 127701000 127705000 127700025
ACRO127700100EA 32.4 EUR
ACRO127700100 ACRO127701000 ACRO127705000 ACRO127700025
4-Methylstyrene 98% stabilised
4-Methylstyrene
Formula: C₉H₁₀
MW: 118,18 g/mol
Boiling Pt: 170…175 °C (760 mmHg)
Melting Pt: –34 °C
Density: 0,896 g/cm³
Flash Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008621
CAS Number: 622-97-9
EINECS: 210-762-8
UN: 2618
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5410 to 1.5430 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.1 % TBC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR