Methyl red sodium salt
Supplier: Roth Carl
Synonyms:
Methyl red water soluble, 2-(4-Dimethylaminophenylazo)benzoic acid sodium salt, Sodium 2-(p-(dimethylamino)phenylazo)benzoate
5003.1
5003.2
ROTH5003.1EA
38.6
EUR
ROTH5003.1
ROTH5003.2
Methyl red sodium salt
Methyl red sodium salt
Formula:
C₁₅H₁₄N₃NaO₂ MW: 291,28 g/mol Melting Pt: > 180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002426 CAS Number: 845-10-3 EINECS: 212-682-9 Merck Index: 13,06142 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...