Alginic acid

Supplier: Roth Carl

8696.1 8696.2
ROTH8696.1EA 0 EUR
ROTH8696.1 ROTH8696.2
Alginic acid
Alginic acid
Formula: (C₆H₈O₆)n
Storage Temperature: Ambient
MDL Number: MFCD00081309
CAS Number: 9005-32-7
EINECS: 232-680-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR