Dicopper selenide ≥99.5% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Copper(I) selenide

45660.09 45660.18
45660.09EA 122 EUR
45660.09 45660.18
Dicopper selenide ≥99.5% (metals basis)
Dicopper selenide

Purity: 99.5% CAS No.: 20405-64-5 Molecular Formula: Cu2Se Formula Weight: 206.05 Physical Form: 2mm & down pieces Density: 6.84 Appearance: Dark blue to black Melting Point: 1113° Merck Reference: 14,2666 UN No.: 3077 MDL No.: MFCD00049908

Formula: Cu₂Se
MW: 206.05 g/mol
Melting Pt: 1113
Density: 6.84
Storage Temperature: Ambient
MDL Number: MFCD00049908
CAS Number: 20405-64-5
EINECS: 243-796-7
UN: 3077
ADR: 9 III
Merck Index: 14,2666

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR